Learn More
ACROBiosystems Human CD3 epsilon Protein, Mouse IgG2a Fc Tag
Marca:  ACROBiosystems CDN-H5253-100ug
Encompass_Preferred
Codice prodotto. 17286160
  • 457.25€ / 100µg
Esplora altre offerte speciali
Questo articolo non è restituibile. Consulta la politica di reso
Descrizione
Descrizione
CD3e molecule, epsilon is also known as CD3E, is a T-cell surface single-pass type I membrane glycoprotein. CD3E contains 1 Ig-like (immunoglobulin-like) domain and 1 ITAM domain. CD3E, together with CD3-gamma, CD3-delta and CD3-zeta, and the T-cell receptor alpha/beta and gamma/delta heterodimers, forms the T cell receptor-CD3 complex. This complex plays an important role in coupling antigen recognition to several intracellular signal-transduction pathways. The genes encoding the epsilon, gamma and delta polypeptides are located in the same cluster on chromosome 11. The epsilon polypeptide plays an essential role in T-cell development. CD3E plays an essential role in T-cell development, and defects in CD3E gene cause severe immunodeficiency. CD3E gene has also been linked to a susceptibility to type I diabetes in women. CD3E has been shown to interact with TOP2B, CD3EAP and NCK2.
This protein carries a mouse IgG2a Fc tag at the C-terminus. The protein has a calculated MW of 38.6 kDa.
Specifica
Specifica
NP_000724.1
100 μg
Asp 23 - Asp 126
FLJ18683,T3E,TCRE,CD3E,CD3-epsilon
Unconjugated
Mouse IgG Fc Tag
Powder
38.6 kDa
Human CD3 epsilon, Mouse IgG2a Fc Tag (CDN-H5253) is expressed from human 293 cells (HEK293). It contains AA Asp 23 - Asp 126 (Accession # NP_000724.1).
-20°C
CD3 epsilon
Human
HEK293
Bio-Markers & CD Antigens