Learn More
ACROBiosystems Human CD34 Protein, His Tag
Marca:  ACROBiosystems CD4-H52H9-100ug
Dettagli aggiuntivi : Peso : 0.00010kg
Codice prodotto. 17286100
  • 522.57€ / 100µg
Spedizione stimata: 15-05-2024
per visualizzare lo stock disponibile
Descrizione
Descrizione
CD34 molecule is a cluster of differentiation molecule present on certain cells within the human body. It is a cell surface glycoprotein and functions as a cell-cell adhesion factor. It may also mediate the attachment of stem cells to bone marrow extracellular matrix or directly to stromal cells. As a ∼ 110 kDa monomeric cell surface antigen, CD34 is highly glycosylated with nine potential N-linked and numerous potential O-linked glycosylation sites in its extracellular domain. The CD34 protein is a member of a family of single-pass transmembrane sialomucin proteins that show expression on early hematopoietic and vascular-associated tissue. CD34 is also an important adhesion molecule and is required for T cells to enter lymph nodes. It is expressed on lymph node endothelia whereas the L-selectin to which it binds is on the T cell. It was indicated that CD34 is a phosphorylation target for activated PKC, and couples to the hematopoietic adapter protein CrkL, which were involved in CD34 signaling pathways. CD34 is abberantly expressed in many kinds of tumors and is implicated in leukemogenesis.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 29.4 kDa. The protein migrates as 67-96 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Specifica
Specifica
NP_001020280.1
100 μg
Ser 32 - Thr 290
CD34,RP11-328D5.2
Unconjugated
His Tag
Less than 1.0 EU / μg by the LAL method.
Powder
>90% as determined by SDS-PAGE.
29.4 kDa
Human CD34, His Tag (CD4-H52H9) is expressed from human 293 cells (HEK293). It contains AA Ser 32 - Thr 290 (Accession # P28906-1).
-20°C
CD34
Human
PBS, pH7.4
HEK293
Bio-Markers & CD Antigens