missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human CEP112 Full-length ORF (NP_001032402.1, 1 a.a. - 211 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00201134-P01.10ug

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16129263

  • 349.00€ / 10µg
Spedizione stimata: 07-11-2023
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

This gene encodes a protein with filament, myosin tail and ATPase domains. Orthologs of this gene exist in mouse, rat and chimp. Alternate transcriptional splice variants, encoding different isoforms, have been characterized. [provided by RefSeq]

Sequence: MWASLSLDHPSAKENQALRLIEMREENGNVPKTEQAGSLKPLRDTGKSNLKEKKANSKLKQIEKEYTQKLAKSSQIIAELQTTISSLKEENSQQQLAAERRLQDVRQKFEDEKKQLIRDNDQAIKVLQDELENRSNQVRCAEKKLQHKELESQEQITYIRQEYETKLKGLMPASLRQELEDTISSLKSQVNFLQKRASILQEELTTYQGRR
Specifica

Specifica

NP_001032402.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
50.9kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CCDC46/MACOCO
CEP112
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
201134
CEP112 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MWASLSLDHPSAKENQALRLIEMREENGNVPKTEQAGSLKPLRDTGKSNLKEKKANSKLKQIEKEYTQKLAKSSQIIAELQTTISSLKEENSQQQLAAERRLQDVRQKFEDEKKQLIRDNDQAIKVLQDELENRSNQVRCAEKKLQHKELESQEQITYIRQEYETKLKGLMPASLRQELEDTISSLKSQVNFLQKRASILQEELTTYQGRR
RUO
CEP112
Wheat Germ (in vitro)
GST
Liquid
Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali