missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human CHAF1B Full-length ORF (NP_005432.1, 1 a.a. - 559 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00008208-P01.25ug

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16124688

  • 528.00€ / 25µg
Spedizione stimata: 03-11-2023
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

Chromatin assembly factor I (CAF-I) is required for the assembly of histone octamers onto newly-replicated DNA. CAF-I is composed of three protein subunits, p50, p60, and p150. The protein encoded by this gene corresponds to the p60 subunit and is required for chromatin assembly after replication. The encoded protein is differentially phosphorylated in a cell cycle-dependent manner. In addition, it is normally found in the nucleus except during mitosis, when it is released into the cytoplasm. This protein is a member of the WD-repeat HIR1 family and may also be involved in DNA repair. [provided by RefSeq]

Sequence: MKVITCEIAWHNKEPVYSLDFQHGTAGRIHRLASAGVDTNVRIWKVEKGPDGKAIVEFLSNLARHTKAVNVVRFSPTGEILASGGDDAVILLWKVNDNKEPEQIAFQDEDEAQLNKENWTVVKTLRGHLEDVYDICWATDGNLMASASVDNTAIIWDVSKGQKISIFNEHKSYVQGVTWDPLGQYVATLSCDRVLRVYSIQKKRVAFNVSKMLSGIGAEGEARSYRMFHDDSMKSFFRRLSFTPDGSLLLTPAGCVESGENVMNTTYVFSRKNLKRPIAHLPCPGKATLAVRCCPVYFELRPVVETGVELMSLPYRLVFAVASEDSVLLYDTQQSFPFGYVSNIHYHTLSDISWSSDGAFLAISSTDGYCSFVTFEKDELGIPLKEKPVLNMRTPDTAKKTKSQTHRGSSPGPRPVEGTPASRTQDPSSPGTTPPQARQAPAPTVIRDPPSITPAVKSPLPGPSEEKTLQPSSQNTKAHPSRRVTLNTLQAWSKTTPRRINLTPLKTDTPPSSVPTSVISTPSTEEIQSETPGDAQGSPPELKRPRLDENKGGTESLDP
Specifica

Specifica

NP_005432.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
87.9kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CAF-1/CAF-IP60/CAF1/CAF1A/CAF1P60/MPHOSPH7/MPP7
CHAF1B
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
8208
CHAF1B (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MKVITCEIAWHNKEPVYSLDFQHGTAGRIHRLASAGVDTNVRIWKVEKGPDGKAIVEFLSNLARHTKAVNVVRFSPTGEILASGGDDAVILLWKVNDNKEPEQIAFQDEDEAQLNKENWTVVKTLRGHLEDVYDICWATDGNLMASASVDNTAIIWDVSKGQKISIFNEHKSYVQGVTWDPLGQYVATLSCDRVLRVYSIQKKRVAFNVSKMLSGIGAEGEARSYRMFHDDSMKSFFRRLSFTPDGSLLLTPAGCVESGENVMNTTYVFSRKNLKRPIAHLPCPGKATLAVRCCPVYFELRPVVETGVELMSLPYRLVFAVASEDSVLLYDTQQSFPFGYVSNIHYHTLSDISWSSDGAFLAISSTDGYCSFVTFEKDELGIPLKEKPVLNMRTPDTAKKTKSQTHRGSSPGPRPVEGTPASRTQDPSSPGTTPPQARQAPAPTVIRDPPSITPAVKSPLPGPSEEKTLQPSSQNTKAHPSRRVTLNTLQAWSKTTPRRINLTPLKTDTPPSSVPTSVISTPSTEEIQSETPGDAQGSPPELKRPRLDENKGGTESLDP
RUO
CHAF1B
Wheat Germ (in vitro)
GST
Liquid
Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali