Learn More
Abnova™ Human CHST11 Partial ORF (NP_060883, 230 a.a. - 337 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca:  Abnova™ H00050515-Q01.25ug
Dettagli aggiuntivi : Peso : 0.00010kg
Codice prodotto. 16126386
  • 528.00€ / 25µg
Spedizione stimata: 02-11-2023
per visualizzare lo stock disponibile
Descrizione
Descrizione
Chondroitin 4-sulfotransferases, such as CHST11, catalyze the transfer of sulfate from 3-prime-phosphoadenosine 5-prime-phosphosulfate to position 4 of N-acetylgalactosamine residues in chondroitin (Yamauchi et al., 2000 [PubMed 10722746]).[supplied by OMIM]
Sequence: RKGDDVKFEEFVAYLIDPHTQREEPFNEHWQTVYSLCHPCHIHYDLVGKYETLEEDSNYVLQLAGVGSYLKFPTYAKSTRTTDEMTTEFFQNISSEHQTQLYEVYKLD
Specifica
Specifica
NP_060883
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.62kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
RKGDDVKFEEFVAYLIDPHTQREEPFNEHWQTVYSLCHPCHIHYDLVGKYETLEEDSNYVLQLAGVGSYLKFPTYAKSTRTTDEMTTEFFQNISSEHQTQLYEVYKLD
RUO
CHST11
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
50515
CHST11 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
C4ST/C4ST-1/C4ST1/HSA269537
CHST11
Recombinant
wheat germ expression system