Learn More
Abnova™ Human CIB2 Partial ORF (NP_006374, 1 a.a. - 99 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00010518-Q01.10ug
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
The amino acid sequence the protein encoded by this gene is similar to that of KIP/CIB, calcineurin B, and calmodulin. This suggests that the encoded protein may be a Ca2+-binding regulatory protein that interacts with DNA-dependent protein kinase catalytic subunit (DNA-PKcs). [provided by RefSeq]
Sequence: MGNKQTIFTEEQLDNYQDCTFFNKKDILKLHSRFYELAPNLVPMDYRKSPIVHVPMSLIIQMPELRENPFKERIVAAFSEDGEGNLTFNDFVDMFSVLCSpecifica
NP_006374 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MGNKQTIFTEEQLDNYQDCTFFNKKDILKLHSRFYELAPNLVPMDYRKSPIVHVPMSLIIQMPELRENPFKERIVAAFSEDGEGNLTFNDFVDMFSVLC | |
RUO | |
CIB2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
10518 | |
CIB2 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
KIP2 | |
CIB2 | |
Recombinant | |
wheat germ expression system |