missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human CMAS Full-length ORF (AAH16609.1, 1 a.a. - 263 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00055907-P01.10ug

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16171953

  • 349.00€ / 10µg
Spedizione stimata: 07-11-2023
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

The enzyme encoded by this gene catalyzes the activation of Neu5Ac to Cytidine 5-prime-monophosphate N-acetylneuraminic acid (CMP-Neu5Ac), which provides the substrate required for the addition of sialic acid. Sialic acids of cell surface glycoproteins and glycolipids play a pivotal role in the structure and function of animal tissues. The pattern of cell surface sialylation is highly regulated during embryonic development, and changes with stages of differentiation. Studies of a similar murine protein suggest that this protein localizes to the nucleus. [provided by RefSeq]

Sequence: MDSVEKGAATSVSNPRGRPSRGRPPKLQRNSRGGQGRGVEKPPHLAALILARGGSKGIPLKNIKHLAGVPLIGWVLRAALDSGAFQSVWVSTDHDEIENVAKQFGAQVHRRSSEVSKDSSTSLDAIIEFLNYHNEVDIVGNIQATSPCLHPTDLQKVAEMIREEGYDSVFSVVRRHQFRWSEIQKGVREVTEPLNLNPAKRPRRQDWDGELYENGSFYFAKRHLIEMGYLQGGKMAYYEMRAEHSVDIDVDIDWPIAEQRVLR
Specifica

Specifica

AAH16609.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
56kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CMAS
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, Protein Array, ELISA, Western Blot
55907
CMAS (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MDSVEKGAATSVSNPRGRPSRGRPPKLQRNSRGGQGRGVEKPPHLAALILARGGSKGIPLKNIKHLAGVPLIGWVLRAALDSGAFQSVWVSTDHDEIENVAKQFGAQVHRRSSEVSKDSSTSLDAIIEFLNYHNEVDIVGNIQATSPCLHPTDLQKVAEMIREEGYDSVFSVVRRHQFRWSEIQKGVREVTEPLNLNPAKRPRRQDWDGELYENGSFYFAKRHLIEMGYLQGGKMAYYEMRAEHSVDIDVDIDWPIAEQRVLR
RUO
CMAS
Recombinant
wheat germ expression system
Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali