missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human cMET ProQuantum Immunoassay Kit
The ProQuantum Human cMET Immunoassay Kit is designed to provide quantitative measurements of human cMET in small sample volumes.
748.00€
Specifica
Numero di accesso | AI838057; AUTS9; bHLHe59; CG1705; CG1705 gene product from transcript CG1705-RB; CG1705-PA; CG1705-PB; cMet; c-met; DFNB97; Dmel\CG1705; Dmel_CG1705; DmMet; EC 2.7.10.1; Hepatocyte growth factor receptor; HGF; HGF receptor; HGF receptor c-Met; HGF/SF receptor; Hgfr; HGF-SF receptor; juvenile hormone resistance; Met; met proto-oncogene; met proto-oncogene (hepatocyte growth factor receptor); met proto-oncogene tyrosine kinase; MET proto-oncogene, receptor tyrosine kinase; Met/Met1; methoprene tolerant; Methoprene-tolerant; Methoprene-tolerant protein; Met-PA; Met-PB; Mett; oncogene MET; Par4; proto-oncogene c-Met; RCCP2; resistance (1) juvenile H; resistance to juvenile hormone; Rst(1)JH; sc MET; Scatter factor receptor; SF receptor; soluble c met; Tyrosine-protein kinase Met |
---|---|
Intervallo saggio | 0.32 to 25,000 pg/mL |
Assay Sensitivity | <0.3 pg/mL |
Product Type | ELISA |
Da utilizzare con (apparecchiatura) | qPCR Instruments |
Codice del prodotto | Marca | Quantità | Prezzo | Quantità e disponibilità | |||||
---|---|---|---|---|---|---|---|---|---|
Codice del prodotto | Marca | Quantità | Prezzo | Quantità e disponibilità | |||||
17104303
|
Invitrogen™
A46338 |
96 Tests |
748.00€
pezzi 96 |
Spedizione stimata: 06-06-2024
Eseguire il login per visualizzare lo stock disponibile |
|||||
Descrizione
Utilizing proximity-based amplification technology, the assay combines the analyte specificity of high-affinity antibody-antigen binding with the signal detection and amplification capabilities of real-time PCR to achieve a simple yet powerful next-generation protein quantitation platform. A user-friendly workflow combined with intuitive software for analytics enables sample-to-answer in just 2 hours.
- High sensitivity-detect low levels of protein with greater sensitivity than traditional methods
- Broad dynamic range-5 logarithmic units, minimizing sample dilutions to ensure they fall within the range
- Small sample consumption-use 2–5 μL of sample (compared to 75 μL for triplicate wells with other methods)
- Fast, easy workflow-2 hours from sample to answer, with no wash steps
- No proprietary instrument to purchase-runs on any real-time PCR instrument
Specifica
AI838057; AUTS9; bHLHe59; CG1705; CG1705 gene product from transcript CG1705-RB; CG1705-PA; CG1705-PB; cMet; c-met; DFNB97; Dmel\CG1705; Dmel_CG1705; DmMet; EC 2.7.10.1; Hepatocyte growth factor receptor; HGF; HGF receptor; HGF receptor c-Met; HGF/SF receptor; Hgfr; HGF-SF receptor; juvenile hormone resistance; Met; met proto-oncogene; met proto-oncogene (hepatocyte growth factor receptor); met proto-oncogene tyrosine kinase; MET proto-oncogene, receptor tyrosine kinase; Met/Met1; methoprene tolerant; Methoprene-tolerant; Methoprene-tolerant protein; Met-PA; Met-PB; Mett; oncogene MET; Par4; proto-oncogene c-Met; RCCP2; resistance (1) juvenile H; resistance to juvenile hormone; Rst(1)JH; sc MET; Scatter factor receptor; SF receptor; soluble c met; Tyrosine-protein kinase Met | |
<0.3 pg/mL | |
qPCR Instruments | |
MET | |
Research Use Only | |
c-Met | |
2 hr |
0.32 to 25,000 pg/mL | |
ELISA | |
P08581 | |
Assay Dilution Buffer Antibody-Conjugate A Antibody-Conjugate B Antibody-Conjugate Dilution Buffer Standard Master Mix Ligase | |
-20°C | |
30 min |