missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human CMTM8 Full-length ORF (NP_849199.2, 1 a.a. - 173 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00152189-P02.10ug

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16128723

  • 349.00€ / 10µg
Spedizione stimata: 03-11-2023
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

This gene belongs to the chemokine-like factor gene superfamily, a novel family that is similar to the chemokine and the transmembrane 4 superfamilies. This gene is one of several chemokine-like factor genes located in a cluster on chromosome 3. This gene is widely expressed in many tissues, but the exact function of the encoded protein is unknown. [provided by RefSeq]

Sequence: MEEPQRARSHTVTTTASSFAENFSTSSSSFAYDREFLRTLPGFLIVAEIVLGLLVWTLIAGTEYFRVPAFGWVMFVAVFYWVLTVFFLIIYITMTYTRIPQVPWTTVGLCFNGSAFVLYLSAAVVDASSVSPERDSHNFNSWAASSFFAFLVTICYAGNTYFSFIAWRSRTIQ
Specifica

Specifica

NP_849199.2
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
46kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CKLFSF8/CKLFSF8-V2
CMTM8
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
152189
CMTM8 (Human) Recombinant Protein (P02)
12.5% SDS-PAGE Stained with Coomassie Blue.
MEEPQRARSHTVTTTASSFAENFSTSSSSFAYDREFLRTLPGFLIVAEIVLGLLVWTLIAGTEYFRVPAFGWVMFVAVFYWVLTVFFLIIYITMTYTRIPQVPWTTVGLCFNGSAFVLYLSAAVVDASSVSPERDSHNFNSWAASSFFAFLVTICYAGNTYFSFIAWRSRTIQ
RUO
CMTM8
Wheat Germ (in vitro)
GST
Liquid
Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali