Learn More
Abnova™ Human CRTAM Full-length ORF (AAH70266.1, 1 a.a. - 393 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00056253-P01.25ug
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
The CRTAM gene is upregulated in CD4 (see MIM 186940)-positive and CD8 (see CD8A; MIM 186910)-positive T cells and encodes a type I transmembrane protein with V and C1-like Ig domains (Yeh et al., 2008 [PubMed 18329370]).[supplied by OMIM]
Sequence: MWWRVLSLLAWFPLQEASLTNHTETITVEEGQTLTLKCVTSLRKNSSLQWLTPSGFTIFLNEYPVLKNSKYQLLHHSANQLSITVPNVTLQDEGVYKCLHYSDSVSTKEVKVIVLATPFKPILEASVIRKQNGEEHVVLMCSTMRSKPPPQITWLLGNSMEVSGGTLHEFETDGKKCNTTSTLIIHTYGKNSTVDCIIRHRGLQGRKLVAPFRFEDLVTDEETASDALERNSLSSQDPQQPTSTVSVTEDSSTSEIDKEEKEQTTQDPDLTTEANPQYLGLARKKSGILLLTLVSFLIFILFIIVQLFIMKLRKTHVIWKKENEVSEHTLESYRSRSNNEETSSEEKNGQSSHPMRCMNYITKLYSEGKTKRKENVQHSKLEEKHIQVPESIVSpecifica
AAH70266.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
71.1kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CRTAM | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
56253 | |
CRTAM (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MWWRVLSLLAWFPLQEASLTNHTETITVEEGQTLTLKCVTSLRKNSSLQWLTPSGFTIFLNEYPVLKNSKYQLLHHSANQLSITVPNVTLQDEGVYKCLHYSDSVSTKEVKVIVLATPFKPILEASVIRKQNGEEHVVLMCSTMRSKPPPQITWLLGNSMEVSGGTLHEFETDGKKCNTTSTLIIHTYGKNSTVDCIIRHRGLQGRKLVAPFRFEDLVTDEETASDALERNSLSSQDPQQPTSTVSVTEDSSTSEIDKEEKEQTTQDPDLTTEANPQYLGLARKKSGILLLTLVSFLIFILFIIVQLFIMKLRKTHVIWKKENEVSEHTLESYRSRSNNEETSSEEKNGQSSHPMRCMNYITKLYSEGKTKRKENVQHSKLEEKHIQVPESIV | |
RUO | |
CRTAM | |
Recombinant | |
wheat germ expression system |