missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human DFNB31 Partial ORF (NP_056219, 808 a.a. - 907 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00025861-Q01.25ug
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
In rat brain, Cip98 interacts with a calmodulin-dependent serine kinase, CASK (MIM 300172), and may be involved in the formation of scaffolding protein complexes that facilitate synaptic transmission in the central nervous system (CNS) (Yap et al., 2003 [PubMed 12641734]). Mutations in this gene, also known as WHRN, cause DFNB31 (MIM 607084).[supplied by OMIM]
Sequence: GLLEPTSTLVRVKKSAATLGIAIEGGANTRQPLPRIVTIQRGGSAHNCGQLKVGHVILEVNGLTLRGKEHREAARIIAEAFKTKDRDYIDFLVTEFNVMLSpecifica
NP_056219 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
GLLEPTSTLVRVKKSAATLGIAIEGGANTRQPLPRIVTIQRGGSAHNCGQLKVGHVILEVNGLTLRGKEHREAARIIAEAFKTKDRDYIDFLVTEFNVML | |
RUO | |
DFNB31 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
25861 | |
DFNB31 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CIP98/DKFZp434N014/KIAA1526/RP11-9M16.1/USH2D/WHRN/WI | |
DFNB31 | |
Recombinant | |
wheat germ expression system |