missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human DGKH Partial ORF (AAH43292, 831 a.a. - 930 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00160851-Q01.10ug

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16172237

  • 349.00€ / 10µg
Spedizione stimata: 03-11-2023
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

This gene encodes a member of the diacylglycerol kinase (DGK) enzyme family of proteins, specifically the type II DGK subfamily. Members of this family are involved in regulating the intracellular concentrations of diacylglycerol and phosphatidic acid. Two transcript variants encoding distinct isoforms have been identified for this gene. [provided by RefSeq]

Sequence: SGKPVLRTHLYIHHAIDLATEEVSQMQLCSQAAEELITRICDAATIHCLLEQELAHAVNACSHALNKANPRCPESLTRDTATEIAINVKALYNETESLLV
Specifica

Specifica

AAH43292
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.63kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
SGKPVLRTHLYIHHAIDLATEEVSQMQLCSQAAEELITRICDAATIHCLLEQELAHAVNACSHALNKANPRCPESLTRDTATEIAINVKALYNETESLLV
RUO
DGKH
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
160851
DGKH (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DGKeta/DKFZp761I1510
DGKH
Recombinant
wheat germ expression system
Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali