missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human DHDDS Full-length ORF (AAH04117.1, 1 a.a. - 294 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00079947-P01.25ug

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16124663

  • 528.00€ / 25µg
Spedizione stimata: 07-11-2023
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

Dehydrodolichyl diphosphate (dedol-PP) synthase catalyzes cis-prenyl chain elongation to produce the polyprenyl backbone of dolichol, a glycosyl carrier lipid required for the biosynthesis of several classes of glycoproteins.[supplied by OMIM]

Sequence: MSWIKEGELSLWERFCANIIKAGPMPKHIAFIMDGNRRYAKKCQVERQEGHSQGFNKLAETLRWCLNLGILEVTVYAFSIENFKRSKSEVDGLMDLARQKFSRLMEEKCFLNVCFAYTSRHEISNAVREMAWGVEQGLLDPSDISESLLDKCLYTNRSPHPDILIRTSGEVRLSDFLLWQTSHSCLVFQPVLWPEYTFWNLFEAILQFQMNHSVLQKARDMYAEERKRQQLERDQATVTEQLLREGLQASGDAQLRRTRLHKLSARREERVQGFLQALELKRADWLARLGTASA
Specifica

Specifica

AAH04117.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
60.7kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CIT/CPT/DS/FLJ13102/HDS
DHDDS
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
79947
DHDDS (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MSWIKEGELSLWERFCANIIKAGPMPKHIAFIMDGNRRYAKKCQVERQEGHSQGFNKLAETLRWCLNLGILEVTVYAFSIENFKRSKSEVDGLMDLARQKFSRLMEEKCFLNVCFAYTSRHEISNAVREMAWGVEQGLLDPSDISESLLDKCLYTNRSPHPDILIRTSGEVRLSDFLLWQTSHSCLVFQPVLWPEYTFWNLFEAILQFQMNHSVLQKARDMYAEERKRQQLERDQATVTEQLLREGLQASGDAQLRRTRLHKLSARREERVQGFLQALELKRADWLARLGTASA
RUO
DHDDS
Wheat Germ (in vitro)
GST
Liquid
Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali