missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human DMC1 Partial ORF (NP_008999, 237 a.a. - 339 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00011144-Q01.25ug

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16126252

  • 528.00€ / 25µg
Spedizione stimata: 02-11-2023
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

The protein encoded by this gene is essential for meiotic homologous recombination. Genetic recombination in meiosis plays an important role in generating diversity of genetic information. The product of this gene is structurally and evolutionary related to the products of the yeast RAD51 and E. coli RecA genes. Alternative splice variants of this gene have been described but their full-length nature has not been determined. [provided by RefSeq]

Sequence: GELAERQQKLAQMLSRLQKISEEYNVAVFVTNQMTADPGATMTFQADPKKPIGGHILAHASTTRISLRKGRGELRIAKIYDSPEMPENEATFAITAGGIGDAK
Specifica

Specifica

NP_008999
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.07kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
GELAERQQKLAQMLSRLQKISEEYNVAVFVTNQMTADPGATMTFQADPKKPIGGHILAHASTTRISLRKGRGELRIAKIYDSPEMPENEATFAITAGGIGDAK
RUO
DMC1
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
11144
DMC1 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DMC1H/HsLim15/LIM15/MGC150472/MGC150473/dJ199H16.1
DMC1
Recombinant
wheat germ expression system
Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali