missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human DOHH Full-length ORF (NP_112594.1, 1 a.a. - 302 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00083475-P01.10ug

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16125353

  • 349.00€ / 10µg
Spedizione stimata: 26-10-2023
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

This gene encodes a metalloenzyme that catalyzes the last step in the conversion of lysine to the unique amino acid hypusine in eukaryotic initiation factor 5A. The encoded protein hydroxylates deoxyhypusine to form hypusine in the mature eukaryotic initiation factor 5A protein. Alternative splicing results in multiple transcript variants

Sequence: MVTEQEVDAIGQTLVDPKQPLQARFRALFTLRGLGGPGAIAWISQAFDDDSALLKHELAYCLGQMQDARAIPMLVDVLQDTRQEPMVRHEAGEALGAIGDPEVLEILKQYSSDPVIEVAETCQLAVRRLEWLQQHGGEPAAGPYLSVDPAPPAEERDVGRLREALLDESRPLFERYRAMFALRNAGGEEAALALAEGLHCGSALFRHEVGYVLGQLQHEAAVPQLAAALARCTENPMVRHECAEALGAIARPACLAALQAHADDPERVVRESCEVALDMYEHETGRAFQYADGLEQLRGAPS
Specifica

Specifica

NP_112594.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
59.3kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
HLRC1/MGC4293
DOHH
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
83475
DOHH (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MVTEQEVDAIGQTLVDPKQPLQARFRALFTLRGLGGPGAIAWISQAFDDDSALLKHELAYCLGQMQDARAIPMLVDVLQDTRQEPMVRHEAGEALGAIGDPEVLEILKQYSSDPVIEVAETCQLAVRRLEWLQQHGGEPAAGPYLSVDPAPPAEERDVGRLREALLDESRPLFERYRAMFALRNAGGEEAALALAEGLHCGSALFRHEVGYVLGQLQHEAAVPQLAAALARCTENPMVRHECAEALGAIARPACLAALQAHADDPERVVRESCEVALDMYEHETGRAFQYADGLEQLRGAPS
RUO
DOHH
Wheat Germ (in vitro)
GST
Liquid
Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali