missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human DTX3L Partial ORF (NP_612144, 3 a.a. - 110 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00151636-Q01.10ug
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
Sequence: SHLRPPSPLLVRVYKSGPRVRRKLESYFQSSKSSGGGECTVSTQEHEAPGTFRVEFSERAAKERVLKKGEHQILVDEKPVPIFLVPTENSIKKNTRPQISSLTQSQAESpecifica
NP_612144 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.62kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
SHLRPPSPLLVRVYKSGPRVRRKLESYFQSSKSSGGGECTVSTQEHEAPGTFRVEFSERAAKERVLKKGEHQILVDEKPVPIFLVPTENSIKKNTRPQISSLTQSQAE | |
RUO | |
DTX3L | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
151636 | |
DTX3L (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
BBAP | |
DTX3L | |
Recombinant | |
wheat germ expression system |