missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human DUSP18 Full-length ORF (NP_689724.3, 1 a.a. - 188 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00150290-P01.10ug

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16128603

  • 335.00€ / 10µg
Spedizione stimata: 28-08-2024
per visualizzare lo stock disponibile

Descrizione

Descrizione

DUSP18 is a member of the dual-specificity phosphatase (DSP) family (see DUSP1; MIM 600714), which catalyzes dephosphorylation of phosphotyrosine and phosphothreonine residues (Hood et al., 2002 [PubMed 12408986]).[supplied by OMIM]

Sequence: MTAPSCAFPVQFRQPSVSGLSQITKSLYISNGVAANNKLMLSSNQITMVINVSVEVVNTLYEDIQYMQVPVADSPNSRLCDFFDPIADHIHSVEMKQGRTLLHCAAGVSRSAALCLAYLMKYHAMSLLDAHTWTKSCRPIIRPNSGFWEQLIHYEFQLFGKNTVHMVSSPVGMIPDIYEKEVRLMIPL
Specifica

Specifica

NP_689724.3
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
47.5kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DSP18/DUSP20/LMWDSP20/MGC32658/bK963H5.1
DUSP18
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
150290
DUSP18 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MTAPSCAFPVQFRQPSVSGLSQITKSLYISNGVAANNKLMLSSNQITMVINVSVEVVNTLYEDIQYMQVPVADSPNSRLCDFFDPIADHIHSVEMKQGRTLLHCAAGVSRSAALCLAYLMKYHAMSLLDAHTWTKSCRPIIRPNSGFWEQLIHYEFQLFGKNTVHMVSSPVGMIPDIYEKEVRLMIPL
RUO
DUSP18
Wheat Germ (in vitro)
GST
Liquid
Suggerimenti di prodotto

Suggerimenti di prodotto

Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali