missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human ELOVL4 Partial ORF (NP_073563, 99 a.a. - 154 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00006785-Q01.25ug

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16126025

  • 528.00€ / 25µg
Spedizione stimata: 07-11-2023
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

This gene encodes a membrane-bound protein which is a member of the ELO family, proteins which participate in the biosynthesis of fatty acids. Consistent with the expression of the encoded protein in photoreceptor cells of the retina, mutations and small deletions in this gene are associated with Stargardt-like macular dystrophy (STGD3) and autosomal dominant Stargardt-like macular dystrophy (ADMD), also referred to as autosomal dominant atrophic macular degeneration. [provided by RefSeq]

Sequence: MGSYNAGYSYICQSVDYSNNVHEVRIAAALWWYFVSKGVEYLDTVFFILRKKNNQV
Specifica

Specifica

NP_073563
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
31.9kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
MGSYNAGYSYICQSVDYSNNVHEVRIAAALWWYFVSKGVEYLDTVFFILRKKNNQV
RUO
ELOVL4
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
6785
ELOVL4 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ADMD/FLJ17667/FLJ92876/STGD2/STGD3
ELOVL4
Recombinant
wheat germ expression system
Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali