missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human ENO1P Full-length ORF (AAH46928.1, 1 a.a. - 388 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00002025-P01.10ug

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16124541

  • 349.00€ / 10µg
Spedizione stimata: 07-11-2023
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

Sequence: MSILKIHARELFDSRGNPTVEVDLFTSEGLFRAAVPSGASTGIYEVLELQDNDKTRYMGKGVSKPVEPINKTIAPVLVSKKLNVTEQEKIDKLMIEMDGTENKSKFGANAILGVSLAACKASAVEKGVPLYHHIADLSGNSKVILPVPVFNVINGSSHAVTKLAMQEFMVLPVGAANFREAMPIGAEVYHSLKNVIKEKYGKDATGVGDGGAFAPNILENKEGLELLKTAIGKAGYTDKVIVSMDVEASEFFRSGKYDLEFKFLDDPTRYISPDCLADLYKSFIKNYPVVSTEDPFDQDDWGAWQKFTASAGIQVVEDDLRVTNPKRTASAVNEKKCNCLLLKVNQIRSVTESLQACKLAQANGWCVMVPHHSGETENTFITDLVVGL
Specifica

Specifica

AAH46928.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
68.7kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ENO1P
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, Protein Array, ELISA, Western Blot
2025
ENO1P (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MSILKIHARELFDSRGNPTVEVDLFTSEGLFRAAVPSGASTGIYEVLELQDNDKTRYMGKGVSKPVEPINKTIAPVLVSKKLNVTEQEKIDKLMIEMDGTENKSKFGANAILGVSLAACKASAVEKGVPLYHHIADLSGNSKVILPVPVFNVINGSSHAVTKLAMQEFMVLPVGAANFREAMPIGAEVYHSLKNVIKEKYGKDATGVGDGGAFAPNILENKEGLELLKTAIGKAGYTDKVIVSMDVEASEFFRSGKYDLEFKFLDDPTRYISPDCLADLYKSFIKNYPVVSTEDPFDQDDWGAWQKFTASAGIQVVEDDLRVTNPKRTASAVNEKKCNCLLLKVNQIRSVTESLQACKLAQANGWCVMVPHHSGETENTFITDLVVGL
RUO
ENO1P
Recombinant
wheat germ expression system
Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali