Learn More
ACROBiosystems Human ENPP3 / CD203c Protein, His Tag
Marca:  ACROBiosystems EN3-H52H4-100ug
Dettagli aggiuntivi : Peso : 0.00010kg
 vedi altre versioni di questo prodotto
Codice prodotto. 17106281
  • dietro preventivo / 100µg
Questo articolo richiede un aggiornamento del prezzo prima di essere ordinato.
Richiedi qui il prezzo per questo articolo.
Descrizione
Descrizione
The human NPP family contains seven members which can be classified into two groups according to their substrate preferences. The first class comprises the nucleotide-degrading proteins NPP1, 3 and 4. NPP3 (CD203c, ENPP3) is expressed in multiple organs, including on epithelial and mucosal surfaces, and notably on basophils and mast cells. Activation of basophils by antigen-bound IgE leads to release of inflammatory mediators and rapid upregulation of NPP3 to the cell surface. This protein is in fact a common marker for diagnosing allergen sensitivity with patient basophils by flow cytometry. Basophils and mast cells mediate the response to certain pathogens, as well as acute and chronic allergic reactions. Following activation, these cells release ATP, which further stimulates them in an autocrine manner. NPP3 upregulation serves to degrade ATP and suppress chronic allergic inflammation (but not the acute response).
This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 96.9 kDa. The protein migrates as 115-120 kDa and 121-130 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Specifica
Specifica
NP_005012.2
96.9 kDa
100 µg
Leu 48 - Ile 875
ENPP3,CD203c,NPP3,E-NPP3,PD-Ibeta,NPPase,PDNP3
Unconjugated
50 mM Tris, 150 mM NaCl, pH8.0
HEK293
Human
Powder
>90% as determined by SDS-PAGE
Human ENPP3, His Tag (EN3-H52H4) is expressed from human 293 cells (HEK293). It contains AA Leu 48 - Ile 875 (Accession # O14638-1).
-20°C
ENPP3
His Tag
Less than 1.0 EU / µg by the LAL method.
ADC