missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human EPB41L4A Full-length ORF (AAH31042.1, 1 a.a. - 185 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00064097-P01.10ug

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16133383

  • 349.00€ / 10µg
Spedizione stimata: 26-10-2023
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

Members of the band 4.1 protein superfamily, including EPB41L4A, are thought to regulate the interaction between the cytoskeleton and plasma membrane (Ishiguro et al., 2000 [PubMed 10874211]).[supplied by OMIM]

Sequence: MGCFCAVPEEFYCEVLLLDESKLTLTTQQQGIKKSTKGSVVLDHVFHHVNLVEIDYFGLRYCDRSHQTYWLDPAKTLAEHKELINTGPPYTLYFGIKFYAEDPCKLKEEITRYQFFLQVKQDVLQGRLPCPVNTAAQLGAYAIQSELGDYDPYKHTAGYVSEYRFVPDQKEELEEAIERIHKTLM
Specifica

Specifica

AAH31042.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
47.8kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
EPB41L4/FLJ38738/NBL4
EPB41L4A
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
64097
EPB41L4A (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MGCFCAVPEEFYCEVLLLDESKLTLTTQQQGIKKSTKGSVVLDHVFHHVNLVEIDYFGLRYCDRSHQTYWLDPAKTLAEHKELINTGPPYTLYFGIKFYAEDPCKLKEEITRYQFFLQVKQDVLQGRLPCPVNTAAQLGAYAIQSELGDYDPYKHTAGYVSEYRFVPDQKEELEEAIERIHKTLM
RUO
EPB41L4A
Wheat Germ (in vitro)
GST
Liquid
Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali