Learn More
Abnova™ Human FAM50A Partial ORF (NP_004690, 1 a.a. - 88 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00009130-Q01.25ug
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
This gene belongs to the FAM50 family. The encoded protein is highly conserved in length and sequence across different species. It is a basic protein containing a nuclear localization signal, and may function as a DNA-binding protein or a transcriptional factor. [provided by RefSeq]
Sequence: MAQYKGAASEAGRAMHLMKKREKQREQMEQMKQRIAEENIMKSNIDKKFSAHYDAVEAELKSSTVGLVTLNDMKAKQEALVKEREKQLSpecifica
NP_004690 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
35.42kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MAQYKGAASEAGRAMHLMKKREKQREQMEQMKQRIAEENIMKSNIDKKFSAHYDAVEAELKSSTVGLVTLNDMKAKQEALVKEREKQL | |
RUO | |
FAM50A | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
9130 | |
FAM50A (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
9F/DXS9928E/HXC-26/XAP5 | |
FAM50A | |
Recombinant | |
wheat germ expression system |