missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human FANCF Partial ORF (NP_073562.1, 1 a.a. - 91 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00002188-Q01.10ug

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16127964

  • 349.00€ / 10µg
Spedizione stimata: 07-11-2023
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

The Fanconi anemia complementation group (FANC) currently includes FANCA, FANCB, FANCC, FANCD1 (also called BRCA2), FANCD2, FANCE, FANCF, FANCG, FANCI, FANCJ (also called BRIP1), FANCL, FANCM and FANCN (also called PALB2). The previously defined group FANCH is the same as FANCA. Fanconi anemia is a genetically heterogeneous recessive disorder characterized by cytogenetic instability, hypersensitivity to DNA crosslinking agents, increased chromosomal breakage, and defective DNA repair. The members of the Fanconi anemia complementation group do not share sequence similarity; they are related by their assembly into a common nuclear protein complex. This gene encodes the protein for complementation group F. [provided by RefSeq]

Sequence: MESLLQHLDRFSELLAVSSTTYVSTWDPATVRRALQWARYLRHIHRRFGRHGPIRTALERRLHNQWRQEGGFGRGPVPGLANFQALGHCDV
Specifica

Specifica

NP_073562.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
35.75kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
MESLLQHLDRFSELLAVSSTTYVSTWDPATVRRALQWARYLRHIHRRFGRHGPIRTALERRLHNQWRQEGGFGRGPVPGLANFQALGHCDV
RUO
FANCF
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
2188
FANCF (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FAF/MGC126856
FANCF
Recombinant
wheat germ expression system
Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali