missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human FBXO10 Partial ORF (XP_291314, 863 a.a. - 962 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00026267-Q01.10ug

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16128252

  • 349.00€ / 10µg
Spedizione stimata: 07-11-2023
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

Members of the F-box protein family, such as FBXO10, are characterized by an approximately 40-amino acid F-box motif. SCF complexes, formed by SKP1 (MIM 601434), cullin (see CUL1; MIM 603134), and F-box proteins, act as protein-ubiquitin ligases. F-box proteins interact with SKP1 through the F box, and they interact with ubiquitination targets through other protein interaction domains (Jin et al., 2004 [PubMed 15520277]).[supplied by OMIM]

Sequence: RAKALVQENIIFQGKTSKTIFQQISNNRECIMQNNKFLVFKKKSDTWRLVNPPARPHLENSLRRPSAAHNGQKVTAMATRITARVEGGYHSNRSVFCTIL
Specifica

Specifica

XP_291314
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
RAKALVQENIIFQGKTSKTIFQQISNNRECIMQNNKFLVFKKKSDTWRLVNPPARPHLENSLRRPSAAHNGQKVTAMATRITARVEGGYHSNRSVFCTIL
RUO
FBXO10
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
26267
FBXO10 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FBX10/FLJ41992/MGC149840
FBXO10
Recombinant
wheat germ expression system
Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali