missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human FBXO16 Partial ORF (NP_758954, 1 a.a. - 100 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00157574-Q01.10ug

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16172207

  • 349.00€ / 10µg
Spedizione stimata: 07-11-2023
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

This gene encodes a member of the F-box protein family, members of which are characterized by an approximately 40 amino acid motif, the F-box. The F-box proteins constitute one of the four subunits of ubiquitin protein ligase complex called SCFs (SKP1-cullin-F-box), which function in phosphorylation-dependent ubiquitination. The F-box proteins are divided into three classes: Fbws containing WD-40 domains, Fbls containing leucine-rich repeats, and Fbxs containing either different protein-protein interaction modules or no recognizable motifs. The protein encoded by this gene belongs to the Fbx class. [provided by RefSeq]

Sequence: MMAFAPPKNTDGPKMQTKMSTWTPLNHQLLNDRVFEERRALLGKWFDKWTDSQRRRILTGLLERCSLSQQKFCCRKLQEKIPAEALDFTTKLPRVLSLYI
Specifica

Specifica

NP_758954
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
MMAFAPPKNTDGPKMQTKMSTWTPLNHQLLNDRVFEERRALLGKWFDKWTDSQRRRILTGLLERCSLSQQKFCCRKLQEKIPAEALDFTTKLPRVLSLYI
RUO
FBXO16
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
157574
FBXO16 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FBX16/MGC125923/MGC125924/MGC125925
FBXO16
Recombinant
wheat germ expression system
Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali