Learn More
Abnova™ Human FKBP6 Full-length ORF (AAH36817.1, 1 a.a. - 327 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00008468-P02.25ug
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
The protein encoded by this gene is a member of the immunophilin protein family, which play a role in immunoregulation and basic cellular processes involving protein folding and trafficking. The protein may have cis-trans prolyl isomerase activity, and binds to clathrin heavy chain and heat shock protein 72. This gene is found to be deleted in Williams syndrome, and the orthologous gene in mouse is essential for fertility and homologous pairing in male meiosis. [provided by RefSeq]
Sequence: MGGSALNQGVLEGDDAPGQSLYERLSQRMLDISGDRGVLKDVIREGAGDLVAPDASVLVKYSGYLEHMDRPFDSNYFRKTPRLMKLGEDITLWGMELGLLSMRRGELARFLFKPNYAYGTLGCPPLIPPNTTVLFEIELLDFLDCAESDKFCALSAEQQDQFPLQKVLKVAATEREFGNYLFRQNRFYDAKVRYKRALLLLRRRSAPPEEQHLVEAAKLPVLLNLSFTYLKLDRPTIALCYGEQALIIDQKNAKALFRCGQACLLLTEYQKARDFLVRAQKEQPFNHDINNELKKLASCYRDYVDKEKEMWHRMFAPCGDGSTAGESSpecifica
AAH36817.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
63.6kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FKBP36/MGC87179/PPIase | |
FKBP6 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
8468 | |
FKBP6 (Human) Recombinant Protein (P02) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MGGSALNQGVLEGDDAPGQSLYERLSQRMLDISGDRGVLKDVIREGAGDLVAPDASVLVKYSGYLEHMDRPFDSNYFRKTPRLMKLGEDITLWGMELGLLSMRRGELARFLFKPNYAYGTLGCPPLIPPNTTVLFEIELLDFLDCAESDKFCALSAEQQDQFPLQKVLKVAATEREFGNYLFRQNRFYDAKVRYKRALLLLRRRSAPPEEQHLVEAAKLPVLLNLSFTYLKLDRPTIALCYGEQALIIDQKNAKALFRCGQACLLLTEYQKARDFLVRAQKEQPFNHDINNELKKLASCYRDYVDKEKEMWHRMFAPCGDGSTAGES | |
RUO | |
FKBP6 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |