missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human FOSB Full-length ORF (AAH40197.1, 1 a.a. - 289 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00002354-P01.25ug

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16125121

  • 528.00€ / 25µg
Spedizione stimata: 07-11-2023
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

The Fos gene family consists of 4 members: FOS, FOSB, FOSL1, and FOSL2. These genes encode leucine zipper proteins that can dimerize with proteins of the JUN family, thereby forming the transcription factor complex AP-1. As such, the FOS proteins have been implicated as regulators of cell proliferation, differentiation, and transformation. Alternatively spliced transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]

Sequence: MPGSFVPTVTAITTSQDLQWLVQPTLISSMAQSQGQPLASQPPVVDPYDMPGTSYSTPGMSGYSSGGASGSGGPSTSGTTSGPGPARPARARPRRPREETLTPEEEEKRRVRRERNKLAAAKCRNRRRELTDRLQAETDQLEEEKAELESEIAELQKEKERLEFVLVAHKPGCKIPYEEGPGPGPLAEVRDLPGSAPAKEDGFSWLLPPPPPPPLPFQTSQDAPPNLTASLFTHSEVQVLGDPFPVVNPSYTSSFVLTCPEVSAFAGAQRTSGSDQPSDPLNSPSLLAL
Specifica

Specifica

AAH40197.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
57.4kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
AP-1/DKFZp686C0818/G0S3/GOS3/GOSB/MGC42291
FOSB
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
2354
FOSB (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MPGSFVPTVTAITTSQDLQWLVQPTLISSMAQSQGQPLASQPPVVDPYDMPGTSYSTPGMSGYSSGGASGSGGPSTSGTTSGPGPARPARARPRRPREETLTPEEEEKRRVRRERNKLAAAKCRNRRRELTDRLQAETDQLEEEKAELESEIAELQKEKERLEFVLVAHKPGCKIPYEEGPGPGPLAEVRDLPGSAPAKEDGFSWLLPPPPPPPLPFQTSQDAPPNLTASLFTHSEVQVLGDPFPVVNPSYTSSFVLTCPEVSAFAGAQRTSGSDQPSDPLNSPSLLAL
RUO
FOSB
Wheat Germ (in vitro)
GST
Liquid
Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali