missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human FOXP2 Full-length ORF (AAH18016.1, 1 a.a. - 67 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00093986-P01.25ug

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16126863

  • 528.00€ / 25µg
Spedizione stimata: 03-11-2023
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

This gene encodes an evolutionarily conserved transcription factor expressed in fetal and adult brain. This transcription factor is a member of the forkhead/winged-helix (FOX) family of transcription factors, and contains a FOX DNA-binding domain and a large polyglutamine tract. Members of the FOX family of transcription factors are regulators of embryogenesis. The product of this gene is thought to be required for proper development of speech and language regions of the brain during embryogenesis. Although a point mutation in this gene has been associated with the KE pedigree segregating developmental verbal dyspraxia, no association between mutations in this gene and another speech disorder, autism, has been found. Multiple alternative transcripts encoding different isoforms have been identified. [provided by RefSeq]

Sequence: MMQESATETISNSSMNQNGMSTLSSQLDAGSRDGRSSGDTSSEVSTVELLHLQQQQEDVVSYTQVIC
Specifica

Specifica

AAH18016.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
33.6kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CAGH44/DKFZp686H1726/SPCH1/TNRC10
FOXP2
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
93986
FOXP2 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MMQESATETISNSSMNQNGMSTLSSQLDAGSRDGRSSGDTSSEVSTVELLHLQQQQEDVVSYTQVIC
RUO
FOXP2
Wheat Germ (in vitro)
GST
Liquid
Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali