Learn More
Abnova™ Human FUT5 Partial ORF (NP_002025, 95 a.a. - 164 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00002527-Q01.25ug
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Sequence: SEMVPGAADCNITADSSVYPQADAVIVHHWDIMYNPSANLPPPTRPQGQRWIWFSMESPSNCRHLEALDGSpecifica
NP_002025 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
33.44kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
SEMVPGAADCNITADSSVYPQADAVIVHHWDIMYNPSANLPPPTRPQGQRWIWFSMESPSNCRHLEALDG | |
RUO | |
FUT5 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
2527 | |
FUT5 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FUC-TV | |
FUT5 | |
Recombinant | |
wheat germ expression system |