missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human GALNACT-2 Full-length ORF (AAH30268, 1 a.a. - 542 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00055454-P01.25ug
Weitere Details : Peso : 0.00010kg
Beschreibung
Sequence: MPRRGLILHTRTHWLLLGLALLCSLVLFMYLLECAPQTDGNASLPGVVGENYGKEYYQALLQEQEEHYQTRATSLKRQIAQLKQELQEMSEKMRSLQERRNVGANGIGYQSNKEQAPSDLLEFLHSQIDKAEVSIGAKLPSEYGVIPFESFTLMKVFQLEMGLTRHPEEKPVRKDKRDELVEVIEAGLEVINNPDEDDEQEDEEGPLGEKLIFNENDFVEGYYRTERDKGTQYELFFKKADLTEYRHVTLFRPFGPLMKVKSEMIDITRSIINIIVPLAERTEAFVQFMQNFRDVCIHQDKKIHLTVVYFGKEGLSKVKSILESVTSESNFHNYTLVSLNEEFNRGRGLNVGARAWDKGEVLMFFCDVDIYFSAEFLNSCRLNAEPGKKVFYPVVFSLYNPAIVYANQEVPPPVEQQLVHKKDSGFWRDFGFGMTCQYRSDFLTIGGFDMEVKGWGGEDVHLYRKYLHGDLIVIRTPVPGLFHLWHEKRCADELTPEQYRMCIQSKAMNEAPHSHLGMLVFREEIETHLHKQAYRTNSEAVGSpezifikation
AAH30268 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
85.36kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CSGalNAcT-2/DKFZp686H13226/FLJ43310/GALNACT-2/GALNACT2/MGC40204/PRO0082 | |
CSGALNACT2 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
55454 | |
GALNACT-2 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MPRRGLILHTRTHWLLLGLALLCSLVLFMYLLECAPQTDGNASLPGVVGENYGKEYYQALLQEQEEHYQTRATSLKRQIAQLKQELQEMSEKMRSLQERRNVGANGIGYQSNKEQAPSDLLEFLHSQIDKAEVSIGAKLPSEYGVIPFESFTLMKVFQLEMGLTRHPEEKPVRKDKRDELVEVIEAGLEVINNPDEDDEQEDEEGPLGEKLIFNENDFVEGYYRTERDKGTQYELFFKKADLTEYRHVTLFRPFGPLMKVKSEMIDITRSIINIIVPLAERTEAFVQFMQNFRDVCIHQDKKIHLTVVYFGKEGLSKVKSILESVTSESNFHNYTLVSLNEEFNRGRGLNVGARAWDKGEVLMFFCDVDIYFSAEFLNSCRLNAEPGKKVFYPVVFSLYNPAIVYANQEVPPPVEQQLVHKKDSGFWRDFGFGMTCQYRSDFLTIGGFDMEVKGWGGEDVHLYRKYLHGDLIVIRTPVPGLFHLWHEKRCADELTPEQYRMCIQSKAMNEAPHSHLGMLVFREEIETHLHKQAYRTNSEAVG | |
RUO | |
CSGALNACT2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |