Learn More
Abnova™ Human GBA2 Full-length ORF (NP_065995.1, 1 a.a. - 927 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00057704-P01.25ug
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
This gene encodes a microsomal beta-glucosidase that catalyzes the hydrolysis of bile acid 3-O-glucosides as endogenous compounds. Studies to determine subcellular localization of this protein in the liver indicated that the enzyme was mainly enriched in the microsomal fraction where it appeared to be confined to the endoplasmic reticulum. This putative transmembrane protein is thought to play a role in carbohydrate transport and metabolism. [provided by RefSeq]
Sequence: MGTQDPGNMGTGVPASEQISCAKEDPQVYCPEETGGTKDVQVTDCKSPEDSRPPKETDCCNPEDSGQLMVSYEGKAMGYQVPPFGWRICLAHEFTEKRKPFQANNVSLSNMIKHIGMGLRYLQWWYRKTHVEKKTPFIDMINSVPLRQIYGCPLGGIGGGTITRGWRGQFCRWQLNPGMYQHRTVIADQFTVCLRREGQTVYQQVLSLERPSVLRSWNWGLCGYFAFYHALYPRAWTVYQLPGQNVTLTCRQITPILPHDYQDSSLPVGVFVWDVENEGDEALDVSIMFSMRNGLGGGDDAPGGLWNEPFCLERSGETVRGLLLHHPTLPNPYTMAVAARVTAATTVTHITAFDPDSTGQQVWQDLLQDGQLDSPTGQSTPTQKGVGIAGAVCVSSKLRPRGQCRLEFSLAWDMPRIMFGAKGQVHYRRYTRFFGQDGDAAPALSHYALCRYAEWEERISAWQSPVLDDRSLPAWYKSALFNELYFLADGGTVWLEVLEDSLPEELGRNMCHLRPTLRDYGRFGYLEGQEYRMYNTYDVHFYASFALIMLWPKLELSLQYDMALATLREDLTRRRYLMSGVMAPVKRRNVIPHDIGDPDDEPWLRVNAYLIHDTADWKDLNLKFVLQVYRDYYLTGDQNFLKDMWPVCLAVMESEMKFDKDHDGLIENGGYADQTYDGWVTTGPSAYCGGLWLAAVAVMVQMAALCGAQDIQDKFSSILSRGQEAYERLLWNGRYYNYDSSSRPQSRSVMSDQCAGQWFLKACGLGEGDTEVFPTQHVVRALQTIFELNVQAFAGGAMGAVNGMQPHGVPDKSSVQSDEVWVGVVYGLAATMIQEGLTWEGFQTAEGCYRTVWERLGLAFQTPEAYCQQRVFRSLAYMRPLSIWAMQLALQQQQHKKASWPKVKQGTGLRTGPMFGPKEAMANLSPESpecifica
NP_065995.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
131kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
AD035/DKFZp762K054/KIAA1605/MGC16895 | |
GBA2 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
57704 | |
GBA2 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MGTQDPGNMGTGVPASEQISCAKEDPQVYCPEETGGTKDVQVTDCKSPEDSRPPKETDCCNPEDSGQLMVSYEGKAMGYQVPPFGWRICLAHEFTEKRKPFQANNVSLSNMIKHIGMGLRYLQWWYRKTHVEKKTPFIDMINSVPLRQIYGCPLGGIGGGTITRGWRGQFCRWQLNPGMYQHRTVIADQFTVCLRREGQTVYQQVLSLERPSVLRSWNWGLCGYFAFYHALYPRAWTVYQLPGQNVTLTCRQITPILPHDYQDSSLPVGVFVWDVENEGDEALDVSIMFSMRNGLGGGDDAPGGLWNEPFCLERSGETVRGLLLHHPTLPNPYTMAVAARVTAATTVTHITAFDPDSTGQQVWQDLLQDGQLDSPTGQSTPTQKGVGIAGAVCVSSKLRPRGQCRLEFSLAWDMPRIMFGAKGQVHYRRYTRFFGQDGDAAPALSHYALCRYAEWEERISAWQSPVLDDRSLPAWYKSALFNELYFLADGGTVWLEVLEDSLPEELGRNMCHLRPTLRDYGRFGYLEGQEYRMYNTYDVHFYASFALIMLWPKLELSLQYDMALATLREDLTRRRYLMSGVMAPVKRRNVIPHDIGDPDDEPWLRVNAYLIHDTADWKDLNLKFVLQVYRDYYLTGDQNFLKDMWPVCLAVMESEMKFDKDHDGLIENGGYADQTYDGWVTTGPSAYCGGLWLAAVAVMVQMAALCGAQDIQDKFSSILSRGQEAYERLLWNGRYYNYDSSSRPQSRSVMSDQCAGQWFLKACGLGEGDTEVFPTQHVVRALQTIFELNVQAFAGGAMGAVNGMQPHGVPDKSSVQSDEVWVGVVYGLAATMIQEGLTWEGFQTAEGCYRTVWERLGLAFQTPEAYCQQRVFRSLAYMRPLSIWAMQLALQQQQHKKASWPKVKQGTGLRTGPMFGPKEAMANLSPE | |
RUO | |
GBA2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |