Learn More
Abnova™ Human GPR128 Partial ORF (NP_116176.1, 31 a.a. - 130 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca:  Abnova™ H00084873-Q01.10ug
Dettagli aggiuntivi : Peso : 0.00010kg
Codice prodotto. 16130917
  • 335.00€ / 10µg
Spedizione stimata: 28-08-2024
per visualizzare lo stock disponibile
Descrizione
Descrizione
Sequence: RIVIRIQRGKSTSSSSTPTEFCRNGGTWENGRCICTEEWKGLRCTIANFCENSTYMGFTFARIPVGRYGPSLQTCGKDTPNAGNPMAVRLCSLSLYGEIE
Specifica
Specifica
NP_116176.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.63kDa
12.5% SDS-PAGE Stained with Coomassie Blue
RIVIRIQRGKSTSSSSTPTEFCRNGGTWENGRCICTEEWKGLRCTIANFCENSTYMGFTFARIPVGRYGPSLQTCGKDTPNAGNPMAVRLCSLSLYGEIE
RUO
GPR128
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
84873
GPR128 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ14454/FLJ16382/FLJ29035/MGC142011/MGC163260
GPR128
Recombinant
wheat germ expression system