missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human GRK7 Full-length ORF (AAI52978.1, 1 a.a. - 553 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00131890-P01.25ug

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16127953

  • 528.00€ / 25µg
Spedizione stimata: 02-11-2023
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

This gene encodes a member of the guanine nucleotide-binding protein (G protein)-coupled receptor kinase subfamily of the Ser/Thr protein kinase family. It is specifically expressed in the retina and the encoded protein has been shown to phosphorylate cone opsins and initiate their deactivation. [provided by RefSeq]

Sequence: MVDMGALDNLIANTAYLQARKPSDCDSKELQRRRRSLALPGLQGCAELRQKLSLNFHSLCEQQPIGRRLFRDFLATVPTFRKAATFLEDVQNWELAEEGPTKDSALQGLVATCASAPAPGNPQPFLSQAVATKCQAATTEEERVAAVTLAKAEAMAFLQEQPFKDFVTSAFYDKFLQWKLFEMQPVSDKYFTEFRVLGKGGFGEVCAVQVKNTGKMYACKKLDKKRLKKKGGEKMALLEKEILEKVSSPFIVSLAYAFESKTHLCLVMSLMNGGDLKFHIYNVGTRGLDMSRVIFYSAQIACGMLHLHELGIVYRDMKPENVLLDDLGNCRLSDLGLAVEMKGGKPITQRAGTNGYMAPEILMEKVSYSYPVDWFAMGCSIYEMVAGRTPFKDYKEKVSKEDLKQRTLQDEVKFQHDNFTEEAKDICRLFLAKKPEQRLGSREKSDDPRKHHFFKTINFPRLEAGLIEPPFVPDPSVVYAKDIAEIDDFSEVRGVEFDDKDKQFFKNFATGAVPIAWQEEIIETGLFEELNDPNRPTGCEEGNSSKSGVCLLL
Specifica

Specifica

AAI52978.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
87.78kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
GPRK7
GRK7
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
131890
GRK7 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MVDMGALDNLIANTAYLQARKPSDCDSKELQRRRRSLALPGLQGCAELRQKLSLNFHSLCEQQPIGRRLFRDFLATVPTFRKAATFLEDVQNWELAEEGPTKDSALQGLVATCASAPAPGNPQPFLSQAVATKCQAATTEEERVAAVTLAKAEAMAFLQEQPFKDFVTSAFYDKFLQWKLFEMQPVSDKYFTEFRVLGKGGFGEVCAVQVKNTGKMYACKKLDKKRLKKKGGEKMALLEKEILEKVSSPFIVSLAYAFESKTHLCLVMSLMNGGDLKFHIYNVGTRGLDMSRVIFYSAQIACGMLHLHELGIVYRDMKPENVLLDDLGNCRLSDLGLAVEMKGGKPITQRAGTNGYMAPEILMEKVSYSYPVDWFAMGCSIYEMVAGRTPFKDYKEKVSKEDLKQRTLQDEVKFQHDNFTEEAKDICRLFLAKKPEQRLGSREKSDDPRKHHFFKTINFPRLEAGLIEPPFVPDPSVVYAKDIAEIDDFSEVRGVEFDDKDKQFFKNFATGAVPIAWQEEIIETGLFEELNDPNRPTGCEEGNSSKSGVCLLL
RUO
GRK7
Wheat Germ (in vitro)
GST
Liquid
Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali