missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human GUCA1A Partial ORF (NP_000400, 1 a.a. - 93 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00002978-Q01.10ug

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16129084

  • 349.00€ / 10µg
Spedizione stimata: 30-10-2023
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

This gene plays a role in the recovery of retinal photoreceptors from photobleaching. In the recovery phase, the phototransduction messeneger cGMP is replenished by retinal guanylyl cyclase-1 (GC1). GC1 is activated by decreasing Ca(2+) concentrations following photobleaching. The protein encoded by this gene, guanylyl cyclase activating protein 1 (GCAP1), mediates the sensitivity of GC1 to Ca(2+) concentrations. GCAP1 promotes activity of GC1 at low Ca(2+) concentrations and inhibits GC1 activity at high Ca(2+) concentrations. Mutations in this gene cause autosomal dominant cone dystrophy (COD3); a disease characterized by reduced visual acuity associated with progressive loss of color vision. Mutations in this gene prohibit the inactivation of RetGC1 at high Ca(2+) concentrations; causing the constitutive activation of RetGC1 and, presumably, increased cell death. This gene is expressed in retina and spermatagonia. [provided by RefSeq]

Sequence: MGNVMEGKSVEELSSTECHQWYKKFMTECPSGQLTLYEFRQFFGLKNLSPSASQYVEQMFETFDFNKDGYIDFMEYVAALSLVLKGKVEQKLR
Specifica

Specifica

NP_000400
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
35.97kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
MGNVMEGKSVEELSSTECHQWYKKFMTECPSGQLTLYEFRQFFGLKNLSPSASQYVEQMFETFDFNKDGYIDFMEYVAALSLVLKGKVEQKLR
RUO
GUCA1A
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
2978
GUCA1A (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
COD3/GCAP/GCAP1/GUCA/GUCA1
GUCA1A
Recombinant
wheat germ expression system
Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali