missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human HOXD10 Partial ORF (NP_002139.2, 44 a.a. - 143 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00003236-Q01.10ug

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16129574

  • 349.00€ / 10µg
Spedizione stimata: 02-11-2023
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

This gene is a member of the Abd-B homeobox family and encodes a protein with a homeobox DNA-binding domain. It is included in a cluster of homeobox D genes located on chromosome 2. The encoded nuclear protein functions as a sequence-specific transcription factor that is expressed in the developing limb buds and is involved in differentiation and limb development. Mutations in this gene have been associated with Wilm's tumor and congenital vertical talus (also known as “rocker-bottom foot” deformity or congenital convex pes valgus) and/or a foot deformity resembling that seen in Charcot-Marie-Tooth disease. [provided by RefSeq]

Sequence: GTYGMQTCGLLPSLAKREVNHQNMGMNVHPYIPQVDSWTDPNRSCRIEQPVTQQVPTCSFTTNIKEESNCCMYSDKRNKLISAEVPSYQRLVPESCPVEN
Specifica

Specifica

NP_002139.2
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
GTYGMQTCGLLPSLAKREVNHQNMGMNVHPYIPQVDSWTDPNRSCRIEQPVTQQVPTCSFTTNIKEESNCCMYSDKRNKLISAEVPSYQRLVPESCPVEN
RUO
HOXD10
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
3236
HOXD10 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
HOX4/HOX4D/HOX4E/Hox-4.4
HOXD10
Recombinant
wheat germ expression system
Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali