Learn More
Abnova™ Human HOXD11 Partial ORF (NP_067015, 1 a.a. - 76 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00003237-Q01.10ug
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
This gene belongs to the homeobox family of genes. The homeobox genes encode a highly conserved family of transcription factors that play an important role in morphogenesis in all multicellular organisms. Mammals possess four similar homeobox gene clusters, HOXA, HOXB, HOXC and HOXD, located on different chromosomes, consisting of 9 to 11 genes arranged in tandem. This gene is one of several homeobox HOXD genes located in a cluster on chromosome 2. Deletions that remove the entire HOXD gene cluster or the 5' end of this cluster have been associated with severe limb and genital abnormalities. The product of the mouse Hoxd11 gene plays a role in forelimb morphogenesis. [provided by RefSeq]
Sequence: MNDFDECGQSAASMYLPGCAYYVAPSDFASKPSFLSQPSSCQMTFPYSSNLAPHVQPVREVAFRDYGLERAKWPYRSpecifica
NP_067015 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
34.1kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MNDFDECGQSAASMYLPGCAYYVAPSDFASKPSFLSQPSSCQMTFPYSSNLAPHVQPVREVAFRDYGLERAKWPYR | |
RUO | |
HOXD11 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
3237 | |
HOXD11 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
HOX4/HOX4F | |
HOXD11 | |
Recombinant | |
wheat germ expression system |