missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human IFIT3 Partial ORF (NP_001540.2, 392 a.a. - 490 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00003437-Q01.25ug
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
Sequence: STDKEEIKDQPQNVSENLLPQNAPNYWYLQGLIHKQNGDLLQAAKCYEKELGRLLRDAPSGIGSIFLSASELEDGSEEMGQGAVSSSPRELLSNSEQLNSpecifica
NP_001540.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
STDKEEIKDQPQNVSENLLPQNAPNYWYLQGLIHKQNGDLLQAAKCYEKELGRLLRDAPSGIGSIFLSASELEDGSEEMGQGAVSSSPRELLSNSEQLN | |
RUO | |
IFIT3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
3437 | |
IFIT3 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CIG-49/GARG-49/IFI60/IFIT4/IRG2/ISG60/RIG-G | |
IFIT3 | |
Recombinant | |
wheat germ expression system |