Learn More
ACROBiosystems Human IFN-alpha 2b (K46R) Protein, Fc Tag
Marca:  ACROBiosystems IFB-H5253-1mg
Dettagli aggiuntivi : Peso : 0.00010kg
Codice prodotto. 17256230
  • dietro preventivo / 1mg
Questo articolo richiede un aggiornamento del prezzo prima di essere ordinato.
Richiedi qui il prezzo per questo articolo.
Descrizione
Descrizione
IFN-alpha is a cytokine that has an immunomodulatory function. It plays an important role not only in antiviral activity but also in several physiologic functions, such as activation of dendritic cells and accelerated expression of major histocompatibility complex I and II molecules that may cause increased antigen presentation. Variants of human leukocyte interferon alpha 2 (IFN-alpha 2a, alpha 2b, and alpha 2c) differ from each other by changes in their coding regions at nucleotide positions 137 and 170. IFN-alpha 2b (IFNα-2b) is an important cytokine and used for antiviral and anticancer treatment.
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 45.7 kDa.
Specifica
Specifica
Powder
1 mg
Cys 24 - Glu 188 (K46R)
IFN-alpha 2 b,IFNA2B,IFN-alpha 2 (alpha 2 b),interferon alpha 2 b
Unconjugated
HEK293
Human
45.7 kDa
Human IFN-alpha 2 b, Fc Tag (IFB-H5253) is expressed from human 293 cells (HEK293). It contains AA Cys 24 - Glu 188 (K46R) (Accession # P01563-1 (K46R)).
-20°C
IFN-alpha 2 b
Fc Tag
ActiveMax™ Cytokines