Learn More
Human IFN-γ ELISPOT Pair, BD
Convenient set of unlabelled anti-IFN-γ capture antibody and a biotinylated anti-IFN-γ detection antibody
Marca: BD Biosciences 551873
Dettagli aggiuntivi : Peso : 0.02000kg
Includes: Human IFN-γ ELISPOT Capture Antibody (no azide/low endotoxin), Human IFN-γ ELISPOT Detection Antibody (biotinylated) and a Certificate of Analysis that provides lot-specific optimal reagent concentrations
Specifica
5 x 96-well Plates |