Learn More
ACROBiosystems Human IL-11 R alpha Protein, Fc Tag
Marca:  ACROBiosystems ILR-H5256-1mg
Dettagli aggiuntivi : Peso : 0.00010kg
Codice prodotto. 17226280
  • 3357.52€ / 1mg
Spedizione stimata: 15-05-2024
per visualizzare lo stock disponibile
Descrizione
Descrizione
Receptor for interleukin-11 (IL11). The receptor systems for IL6, LIF, OSM, CNTF, IL11 and CT1 can utilize IL6ST for initiating signal transmission. The IL11/IL11RA/IL6ST complex may be involved in the control of proliferation and/or differentiation of skeletogenic progenitor or other mesenchymal cells (Probable). Essential for the normal development of craniofacial bones and teeth. Restricts suture fusion and tooth number.Soluble form of IL11 receptor (sIL11RA) that acts as an agonist of IL11 activity. The IL11:sIL11RA complex binds to IL6ST/gp130 on cell surfaces and induces signaling also on cells that do not express membrane-bound IL11RA in a process called IL11 trans-signaling.
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 64.2 kDa.
Specifica
Specifica
64.2 kDa
Human IL-11 R alpha, Fc Tag (ILR-H5256) is expressed from human 293 cells (HEK293). It contains AA Ser 24 - Ala 370 (Accession # Q14626-1).
-20°C
IL-11 R alpha
Human
HEK293
Cytokines&Growth; Factors
1 mg
Ser 24 - Ala 370
IL-11 R alpha,IL-11 RA,IL11RA,Interleukin-11 receptor subunit alpha,IL-11 receptor subunit alpha,IL-11 R subunit alpha,IL-11 R-alpha
Unconjugated
Fc Tag
Powder