Learn More
ACROBiosystems Human IL-13 R alpha 2 Protein, Fc Tag
MALS verified
Marca:  ACROBiosystems IL2-H5256-1mg
Dettagli aggiuntivi : Peso : 0.00010kg
 vedi altre versioni di questo prodotto
Codice prodotto. 17116401
  • 2090.29€ / 1mg
Spedizione stimata: 21-05-2024
per visualizzare lo stock disponibile
Descrizione
Descrizione
Interleukin-13 receptor subunit alpha-2 is also known as IL13Rα2, IL13Ra2 cluster of differentiation 213A2, CD213A2, CT19, IL-13R, IL13BP, and is a membrane bound protein that in humans is encoded by the IL13RA2 gene. IL13Rα2 is closely related to IL13Rα1, a subunit of the interleukin-13 receptor complex. This protein binds IL13 with high affinity, but lacks any significant cytoplasmic domain, and does not appear to function as a signal mediator. It is, however able to regulate the effects of both IL13 and IL4, despite the fact it is unable to bind directly to the latter. It is also reported to play a role in the internalization of IL13. IL13Rα2 is a component of the cell surface receptors, however, the majority exists in intracellular pools and in soluble form, and thus plays an opposite role as a potent IL13 antagonist compared with IL13Rα1. It also functions as an inhibitor of IL4-dependent pathway probably through the physical interaction between the short intracellular domain of and cytoplasmic domain of IL13Rα2 and the IL4Rα chain. In spite of the failed STAT signaling function, IL13Rα2 dose induce TGF-beta production and fibrosis. Additionally, IL13Rα2has been reported to be abundantly and specifically overexpressed in glioblastoma multiforme.
This protein carries a human IgG1 Fc tag at the N-terminus. The protein has a calculated MW of 63.5 kDa. The protein migrates as 66-80 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Specifica
Specifica
NP_000631.1
1 mg
Asp 27 - Arg 343
IL13RA2,CD213A2,CT19,IL-13 R,IL13BP
Unconjugated
Fc Tag
Less than 1.0 EU / μg by the LAL method.
Powder
>90% as determined by SDS-PAGE
63.5 kDa
Human IL-13 RA2, Fc Tag (MALS verified) (IL2-H5256) is expressed from human 293 cells (HEK293). It contains AA Asp 27 - Arg 343 (Accession # NP_000631.1).
-20°C
IL-13 R alpha 2
Human
PBS, pH7.4
HEK293
Bio-Markers and CD Antigens