missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human INPP1 Full-length ORF (NP_002185.1, 1 a.a. - 399 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00003628-P02.25ug

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16129367

  • 528.00€ / 25µg
Spedizione stimata: 07-11-2023
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

This gene encodes the enzyme inositol polyphosphate-1-phosphatase, one of the enzymes involved in phosphatidylinositol signaling pathways. This enzyme removes the phosphate group at position 1 of the inositol ring from the polyphosphates inositol 1,4-bisphosphate and inositol 1,3,4-trisphophosphate. [provided by RefSeq]

Sequence: MSDILRELLCVSEKAANIARACRQQEALFQLLIEEKKEGEKNKKFAVDFKTLADVLVQEVIKQNMENKFPGLEKNIFGEESNEFTNDWGEKITLRLCSTEEETAELLSKVLNGNKVASEALARVVHQDVAFTDPTLDSTEINVPQDILGIWVDPIDSTYQYIKGSADIKSNQGIFPCGLQCVTILIGVYDIQTGVPLMGVINQPFVSRDPNTLRWKGQCYWGLSYMGTNMHSLQLTISRRNGSETHTGNTGSEAAFSPSFSAVISTSEKETIKAALSRVCGDRIFGAAGAGYKSLCVVQGLVDIYIFSEDTTFKWDSCAAHAILRAMGGGIVDLKECLERNPETGLDLPQLVYHVENEGAAGVDRWANKGGLIAYRSRKRLETFLSLLVQNLAPAETHT
Specifica

Specifica

NP_002185.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
70.4kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MGC110984
INPP1
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
3628
INPP1 (Human) Recombinant Protein (P02)
12.5% SDS-PAGE Stained with Coomassie Blue.
MSDILRELLCVSEKAANIARACRQQEALFQLLIEEKKEGEKNKKFAVDFKTLADVLVQEVIKQNMENKFPGLEKNIFGEESNEFTNDWGEKITLRLCSTEEETAELLSKVLNGNKVASEALARVVHQDVAFTDPTLDSTEINVPQDILGIWVDPIDSTYQYIKGSADIKSNQGIFPCGLQCVTILIGVYDIQTGVPLMGVINQPFVSRDPNTLRWKGQCYWGLSYMGTNMHSLQLTISRRNGSETHTGNTGSEAAFSPSFSAVISTSEKETIKAALSRVCGDRIFGAAGAGYKSLCVVQGLVDIYIFSEDTTFKWDSCAAHAILRAMGGGIVDLKECLERNPETGLDLPQLVYHVENEGAAGVDRWANKGGLIAYRSRKRLETFLSLLVQNLAPAETHT
RUO
INPP1
Wheat Germ (in vitro)
GST
Liquid
Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali