missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human JRK Full-length ORF (AAH43351.1, 1 a.a. - 568 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00008629-P01.25ug

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16172692

  • 508.00€ / 25µg
Spedizione stimata: 26-08-2024
per visualizzare lo stock disponibile

Descrizione

Descrizione

This gene is the human homolog of the mouse jerky gene. The encoded protein has similarity to several nuclear regulatory proteins, including centromere protein B, suggesting that it might function as a DNA-binding protein. Insertional inactivation of this gene in transgenic mice resulted in epileptic seizures. Childhood Absence Epilepsy (CAE) has been mapped to the same chromosomal location (8q24.3) as this gene, making this gene a strong candidate for CAE. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]

Sequence: MASKPAAGKSRGEKRKRVVLTLKEKIDICTRLEKGESRKALMQEYNVGMSTLYDIRAHKAQLLRFFASSDSNKALEQRRTLHTPKLEHLDRVLYEWFLGKRSEGVPVSGPMLIEKAKDFYEQMQLTEPCVFSGGWLWRFKARHGIKKLDASSEKQSADHQAAEQFCAFFRSLAAEHGLSAEQVYNADETGLFWRCLPNPTPEGGAVPGPKQGKDRLTVLMCANATGSHRLKPLAIGKCSGPRAFKGIQHLPVAYKAQGNAWVDKEIFSDWFHHIFVPSVREHFRTIGLPEDSKAVLLLDSSRAHPQEAELVSSNVFTIFLPASVASLVQPMEQGIRRDFMRNFINPPVPLQGPHARYNMNDAIFSVACAWNAVPSHVFRRAWRKLWPSVAFAEGSSSEEELEAECFPVKPHNKSFAHILELVKEGSSCPGQLRQRQAASWGVAGREAEGGRPPAATSPAEVVWSSEKTPKADQDGRGDPGEGEEVAWEQAAVAFDAVLRFAERQPCFSAQEVGQLRALRAVFRSQQQVRRRRGALGAVVKVEALQEGPGGCGATAQSPLPCSSTAGDN
Specifica

Specifica

AAH43351.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
89.2kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DKFZp686C24207/FLJ45729/JH8
JRK
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
8629
JRK (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MASKPAAGKSRGEKRKRVVLTLKEKIDICTRLEKGESRKALMQEYNVGMSTLYDIRAHKAQLLRFFASSDSNKALEQRRTLHTPKLEHLDRVLYEWFLGKRSEGVPVSGPMLIEKAKDFYEQMQLTEPCVFSGGWLWRFKARHGIKKLDASSEKQSADHQAAEQFCAFFRSLAAEHGLSAEQVYNADETGLFWRCLPNPTPEGGAVPGPKQGKDRLTVLMCANATGSHRLKPLAIGKCSGPRAFKGIQHLPVAYKAQGNAWVDKEIFSDWFHHIFVPSVREHFRTIGLPEDSKAVLLLDSSRAHPQEAELVSSNVFTIFLPASVASLVQPMEQGIRRDFMRNFINPPVPLQGPHARYNMNDAIFSVACAWNAVPSHVFRRAWRKLWPSVAFAEGSSSEEELEAECFPVKPHNKSFAHILELVKEGSSCPGQLRQRQAASWGVAGREAEGGRPPAATSPAEVVWSSEKTPKADQDGRGDPGEGEEVAWEQAAVAFDAVLRFAERQPCFSAQEVGQLRALRAVFRSQQQVRRRRGALGAVVKVEALQEGPGGCGATAQSPLPCSSTAGDN
RUO
JRK
Wheat Germ (in vitro)
GST
Liquid
Suggerimenti di prodotto

Suggerimenti di prodotto

Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali