missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human KRT71 Full-length ORF (AAI03919.1, 1 a.a. - 523 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00112802-P01.10ug

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16126953

  • 349.00€ / 10µg
Spedizione stimata: 30-10-2023
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

Keratins are intermediate filament proteins responsible for the structural integrity of epithelial cells and are subdivided into epithelial keratins and hair keratins. This gene encodes a protein that is expressed in the inner root sheath of hair follicles. The type II keratins are clustered in a region of chromosome 12q13

Sequence: MSRQFTCKSGAAAKGGFSGCSAVLSGGSSSSFRAGSKGLSGGFGSRSLYSLGGVRSLNVASGSGKSGGYGFGRGRASGFAGSMFGSVALGPVCPTVCPPGGIHQVTVNESLLAPLNVELDPEIQKVRAQEREQIKALNNKFASFIDKVRFLEQQNQVLETKWELLQQLDLNNCKNNLEPILEGYISNLRKQLETLSGDRVRLDSELRNVQDVVEDYKKRYEEEINKRTAAENEFVLLKKDVDAAYANKVELQAKVESMDQEIKFFRCLFEAEITQIQSHISDMSVILSMDNNRNLDLDSIIDEVRTQYEEIALKSKAEAEALYQTKFQELQLAAGRHGDDLKNTKNEISELTRLIQRIRSEIENVKKQASNLETAIADAEQRGDNALKDARAKLDELEGALHQAKEELARMLREYQELMSLKLALDMEIATYRKLLESEECRMSGEFPSPVSISIISSTSGGSGYGFRPSMVSGGYVANSSNCISGVCSVRGGEGRSRGSANDYKDTLGKGSSLSAPSKKTSR
Specifica

Specifica

AAI03919.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
83.6kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
K6IRS1/KRT6IRS/KRT6IRS1/MGC119390/MGC119391
KRT71
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
112802
KRT71 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MSRQFTCKSGAAAKGGFSGCSAVLSGGSSSSFRAGSKGLSGGFGSRSLYSLGGVRSLNVASGSGKSGGYGFGRGRASGFAGSMFGSVALGPVCPTVCPPGGIHQVTVNESLLAPLNVELDPEIQKVRAQEREQIKALNNKFASFIDKVRFLEQQNQVLETKWELLQQLDLNNCKNNLEPILEGYISNLRKQLETLSGDRVRLDSELRNVQDVVEDYKKRYEEEINKRTAAENEFVLLKKDVDAAYANKVELQAKVESMDQEIKFFRCLFEAEITQIQSHISDMSVILSMDNNRNLDLDSIIDEVRTQYEEIALKSKAEAEALYQTKFQELQLAAGRHGDDLKNTKNEISELTRLIQRIRSEIENVKKQASNLETAIADAEQRGDNALKDARAKLDELEGALHQAKEELARMLREYQELMSLKLALDMEIATYRKLLESEECRMSGEFPSPVSISIISSTSGGSGYGFRPSMVSGGYVANSSNCISGVCSVRGGEGRSRGSANDYKDTLGKGSSLSAPSKKTSR
RUO
KRT71
Wheat Germ (in vitro)
GST
Liquid
Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali