Learn More
Human LAP (TGF-beta 1) Antibody, R&D Systems™
Mouse Monoclonal Antibody has been used in 8 publications
Marca: R&D Systems MAB246-100
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
LAP (TGF-beta 1) Monoclonal specifically detects LAP (TGF-beta 1) in Human samples. It is validated for Neutralization, Immunocytochemistry.
Specifica
| LAP (TGF-beta 1) | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| TGFB1 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human LAP (TGF-beta 1) | |
| 100 μg | |
| Primary | |
| Detects human LAP in direct ELISAs and Western blots. | |
| Human | |
| Ascites |
| Neutralization, Immunocytochemistry | |
| 27235 | |
| Neutralization 1-5 ug/mL, Immunocytochemistry 3-25 ug/mL | |
| CED, DPD1, LAP, LAP (TGFbeta 1), TGFB, TGFB1, TGFbeta, transforming growth factor beta 1 | |
| Mouse | |
| Protein A or G purified from ascites | |
| RUO | |
| 7040 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Titolo del prodotto
For Research Use Only