missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human LAP (TGF-beta 1) Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 8 publications
Marca: R&D Systems MAB246-500
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
LAP (TGF-beta 1) Monoclonal specifically detects LAP (TGF-beta 1) in Human samples. It is validated for Neutralization, Immunocytochemistry.
Specifica
| LAP (TGF-beta 1) | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| TGFB1 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human LAP (TGF-beta 1) | |
| 500 μg | |
| Primary | |
| Detects human LAP in direct ELISAs and Western blots. | |
| Human | |
| Ascites |
| Neutralization, Immunocytochemistry | |
| 27235 | |
| Neutralization 1-5 ug/mL, Immunocytochemistry 3-25 ug/mL | |
| CED, DPD1, LAP, LAP (TGFbeta 1), TGFB, TGFB1, TGFbeta, transforming growth factor beta 1 | |
| Mouse | |
| Protein A or G purified from ascites | |
| RUO | |
| 7040 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
For Research Use Only
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto