missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human LAPTM5 Partial ORF (NP_006753.1, 207 a.a. - 262 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00007805-Q01.25ug

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16127965

  • 528.00€ / 25µg
Spedizione stimata: 07-11-2023
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

This gene encodes a transmembrane receptor that is associated with lysosomes. The encoded protein, also known as E3 protein, may play a role in hematopoiesis. [provided by RefSeq]

Sequence: KCVWRCYRLIKCMNSVEEKRNSKMLQKVVLPSYEEALSLPSKTPEGGPAPPPYSEV
Specifica

Specifica

NP_006753.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
31.9kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
KCVWRCYRLIKCMNSVEEKRNSKMLQKVVLPSYEEALSLPSKTPEGGPAPPPYSEV
RUO
LAPTM5
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
7805
LAPTM5 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ97251/MGC125860/MGC125861
LAPTM5
Recombinant
wheat germ expression system
Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali