Learn More
Abnova™ Human LASS6 Full-length ORF (AAH30800.2, 1 a.a. - 38 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca:  Abnova™ H00253782-P01.10ug
Dettagli aggiuntivi : Peso : 0.00010kg
Codice prodotto. 16129613
  • 349.00€ / 10µg
Spedizione stimata: 07-11-2023
per visualizzare lo stock disponibile
Descrizione
Descrizione
Sequence: MIEVILSLAQMRRTQNLRERIPTLRQPPMGPVVPTGIS
Specifica
Specifica
AAH30800.2
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
30.7kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CerS6/MGC129949/MGC129950
LASS6
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
253782
LASS6 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MIEVILSLAQMRRTQNLRERIPTLRQPPMGPVVPTGIS
RUO
LASS6
Wheat Germ (in vitro)