Learn More
ACROBiosystems Human latent GDF-8 Protein, His Tag
Marca:  ACROBiosystems GD8-H5243-500ug
Dettagli aggiuntivi : Peso : 0.00010kg
Codice prodotto. 17226220
  • 3488.16€ / 500µg
Spedizione stimata: 30-05-2024
per visualizzare lo stock disponibile
Descrizione
Descrizione
Growth differentiation factor 8 (GDF8), also known as myostatin, is a unique member of the transforming growth factor-β superfamily that is expressed in human granulosa cells and has important roles in regulating a variety of ovarian functions. GDF8 acts as a negative regulator of skeletal muscle growth and differentiation. In addition to the expression in the musculoskeletal system, GDF8 is also expressed in various tissues, including the reproductive system.
This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 42 kDa.
Specifica
Specifica
42 kDa
Human latent GDF-8, His Tag (GD8-H5243) is expressed from human 293 cells (HEK293). It contains AA Asn 24 - Ser 375 (Accession # O14793-1).
-20°C
latent GDF-8
Human
HEK293
Bio-Markers & CD Antigens
500 μg
Asn 24 - Ser 375
latent GDF-8, GDF-8, Myostatin, GDF8, MSTN, Growth/differentiation factor 8
Unconjugated
His Tag
Powder