missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human LCN12 Full-length ORF (AAH41168.1, 1 a.a. - 355 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00286256-P01.10ug

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16130064

  • 349.00€ / 10µg
Spedizione stimata: 03-11-2023
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

Members of the lipocalin family, such as LCN12, have a common structure consisting of an 8-stranded antiparallel beta-barrel that forms a cup-shaped ligand-binding pocket or calyx. Lipocalins generally bind small hydrophobic ligands and transport them to specific cells (Suzuki et al., 2004 [PubMed 15363845]).[supplied by OMIM]

Sequence: MRLLCGLWLWLSLLKVLQAQTPTPLPLPPPMQSFQGNQFQGEWFVLGLAGNSFRPEHRALLNAFTATFELSDDGRFEVWNAMTRGQHCDTWSYVLIPAAQPGQFTVDHGVEPGADREETRVVDSDYTQFALMLSRRHTSRLAVLRISLLGRSWLLPPGTLDQFICLGRAQGLSDDNIVFPDVTGNMVHLQACWAVGTGPAGMSLVDPRGAGPSVYPGSSAPACAQGSPGSWVPVLNPGSEPPPAAPGPLSWATSSHPGSPVPGHLLPPQVPCPGPPPPAPPAPGPLSRPTSSHPGSPVLGYLLPPQVPCPGPSPPSGSPVLGHLLPSPIPAHKELGLIPGGALDLSSLPWVAAPA
Specifica

Specifica

AAH41168.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
63.7kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MGC34753/MGC48935
LCN12
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
286256
LCN12 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MRLLCGLWLWLSLLKVLQAQTPTPLPLPPPMQSFQGNQFQGEWFVLGLAGNSFRPEHRALLNAFTATFELSDDGRFEVWNAMTRGQHCDTWSYVLIPAAQPGQFTVDHGVEPGADREETRVVDSDYTQFALMLSRRHTSRLAVLRISLLGRSWLLPPGTLDQFICLGRAQGLSDDNIVFPDVTGNMVHLQACWAVGTGPAGMSLVDPRGAGPSVYPGSSAPACAQGSPGSWVPVLNPGSEPPPAAPGPLSWATSSHPGSPVPGHLLPPQVPCPGPPPPAPPAPGPLSRPTSSHPGSPVLGYLLPPQVPCPGPSPPSGSPVLGHLLPSPIPAHKELGLIPGGALDLSSLPWVAAPA
RUO
LCN12
Wheat Germ (in vitro)
GST
Liquid
Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali